Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Positive test controls include: Mouse lung, Mouse spleen, Rat spleen.
Immunogen sequence: MPSELEKALS NLIDVYHNYS NIQGNHHALY KNDFKKMVTT ECPQFVQNIN IENLFRELDI NSDNAINFEE FLAMVIKVGV ASHKDSHKE
The protein encoded by this gene is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. This protein may function in the inhibition of casein kinase and as a cytokine. Altered expression of this protein is associated with the disease cystic fibrosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Calgranulin-A; Chemotactic cytokine CP-10; chemotactic S100 protein; Leukocyte L1 complex light chain; Migration inhibitory factor-related protein 8; MRP-8; p8; Pro-inflammatory S100 cytokine; Protein S100-A8; S100 A8; S100 calcium binding protein A8 (calgranulin A); S100 calcium-binding protein A8; S100 calcium-binding protein A8 (calgranulin A)
Gene Aliases: 60B8Ag; AI323541; B8Ag; Caga; CFAg; CP-10; MRP8; p8; S100a8
UniProt ID: (Rat) P50115, (Mouse) P27005
Entrez Gene ID: (Rat) 116547, (Mouse) 20201
Molecular Function:
calmodulin-related
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support