Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 Extended to June 20: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 2
In direct ELISAs and Western blots, 40% cross-reactivity with recombinant human (rh) S100A8 is observed and no cross-reactivity with recombinant mouse (rm) S100A4, rmS100A9, rmS100A10, rmS100A11, rhS100B, or rhS100P is observed.
Reconstitute at 0.5 mg/mL in sterile PBS.
The protein encoded by this gene is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. This protein may function in the inhibition of casein kinase and as a cytokine. Altered expression of this protein is associated with the disease cystic fibrosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Calgranulin-A; Chemotactic cytokine CP-10; chemotactic S100 protein; Leukocyte L1 complex light chain; Migration inhibitory factor-related protein 8; MRP-8; Pro-inflammatory S100 cytokine; Protein S100-A8; S100 A8; S100 calcium-binding protein A8
Gene Aliases: 60B8Ag; AI323541; B8Ag; Caga; CFAg; CP-10; MRP8; p8; S100a8
UniProt ID: (Mouse) P27005
Entrez Gene ID: (Mouse) 20201
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support