Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Immunogen sequence: MTCKMSQLE RNIETIINTF HQYSVKLGHP DTLNQGEFKE LVRKDLQNFL KKENKNEKVI EHIMEDLDTN ADKQLSFEEF IMLMARLTWA SHEKMHEGDE GPGHHHKPGL GEGTP (1-114 aa encoded by BC047681)
S100A9 (MRP-14) is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. This protein may function in the inhibition of casein kinase and altered expression of this protein is associated with the disease cystic fibrosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BM-009; calgranulin B; Calgranulin-B; Calprotectin L1H subunit; DKFZp686B04128; intracellular calcium-binding protein (MRP14); Leukocyte L1 complex heavy chain; Macrophage; Migration inhibitory factor-related protein 14; Monocyte; MRP-14; MRP15; MRP16; Myeloid-related protein 14; p14; Protein S100-A9; S100 A9; S100 calcium binding protein A9 (calgranulin B); S100 calcium-binding protein A9; S100 calcium-binding protein A9 (calgranulin B)
Gene Aliases: 60B8AG; AW546964; BEE22; CAGB; CFAG; CGLB; GAGB; L1AG; LIAG; MAC387; MIF; MRP14; NIF; P14; S100A9
UniProt ID: (Human) P06702, (Mouse) P31725, (Rat) P50116
Entrez Gene ID: (Human) 6280, (Mouse) 20202, (Rat) 94195
Molecular Function:
calmodulin-related
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support