Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) S100A10 or rhS100B is observed.
Reconstitute at 0.5 mg/mL in sterile PBS.
The protein encoded by this gene is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21; however, this gene is located at 4p16. This protein, in addition to binding Ca2+, also binds Zn2+ and Mg2+. This protein may play a role in the etiology of prostate cancer.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MIG9; Migration-inducing gene 9 protein; Protein S100-E; Protein S100-P; S100 calcium-binding protein P
Gene Aliases: MIG9; S100E; S100P
UniProt ID: (Human) P25815
Entrez Gene ID: (Human) 6286
Molecular Function: calmodulin-related
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support