Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MRLFTGIVFC SLVMGVTSES WRSFFKEALQ GVGDMGRAYW DIMISNHQNS NRYLYARGNY DAAQRGPGGV WAAKLISRSR VYLQGLIDYY LFGNSSTVLE DSKSNEKAEE WGRSGKDPDR FRPDGLPKKY
SAA4 is a major acute phase reactant. It is an Apolipoprotein of the HDL complex.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C-SAA; Constitutively expressed serum amyloid A protein; Serum amyloid A-4 protein
Gene Aliases: C-SAA; CSAA; SAA4
UniProt ID: (Human) P35542
Entrez Gene ID: (Human) 6291
Molecular Function: apolipoprotein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support