Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Antibody detects endogenous levels of total SACM1L.
Phosphoinositide phosphatase that hydrolyzes PtdIns(3)P and PtdIns(4)P. Has low activity towards PtdIns(3,5)P2.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: phosphatidylinositide phosphatase SAC1; Phosphatidylinositol-3-phosphatase SAC1; Phosphatidylinositol-4-phosphate phosphatase; polyphosphatidylinositol phosphatase; suppressor of actin 1; Suppressor of actin mutations 1-like protein
Gene Aliases: KIAA0851; mKIAA0851; SAC1; Sac1p; SACM1L
UniProt ID: (Human) Q9NTJ5, (Mouse) Q9EP69, (Rat) Q9ES21
Entrez Gene ID: (Human) 22908, (Mouse) 83493, (Rat) 116482
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support