Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSRRKQAKP QHINSEEDQG EQQPQQQTPE FADAAPAAPA AGELGAPVNH PGNDEVASED EATVKRLRRE ETHVCEKCCA EFFSISEFLE HKKNCTKNPP VLIMNDSEGP VPSEDFSGAV LSHQPTSPGS KDCHRENGGS SEDMKEKPDA ESVVYLKTET ALPPTPQDIS YLAKGKVANT NVTLQALRGT KVAVNQRSAD ALPAPVPGAN SIPWVLEQIL CLQQQQLQQI QLTEQIRIQV NMWASHALHS SGAGADTLKT LGSHMSQQVS AAVALLSQKA GSQGLSLDAL KQAKLPHANI PSATSSLSPG LAPFTLKPDG TRVLPNVMSR LPSALLPQAP GSVLFQSPFS T (1-350 aa encoded by BC111714)
Sall4 protein is encoded by the Sall4 gene located on chromosome 20 in humans and belongs to the Spalt-like protein family. It plays an important role in somatic stem cell maintenance and is studied as a stem cell biomarker. It is a zinc finger transcription factor that is upregulated during development and shown to interact with Oct4 and Sox2. It also forms a complex with Nanog to maintain the pluripotency of Embryonic Stem cells (ESCs). Several current research studies focus on its function as an oncogene and tumor biomarker. Some of the reported target genes of Sall4 include p53, Bmi-1, Bcl2 and XIAP.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dJ1112F19.1; Sal-like protein 4; spalt-like transcription factor 4; Zinc finger protein 797; Zinc finger protein SALL4
Gene Aliases: DRRS; HSAL4; SALL4; ZNF797
UniProt ID: (Human) Q9UJQ4
Entrez Gene ID: (Human) 57167
Molecular Function: C2H2 zinc finger transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support