Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This gene encodes subunit 2 of the splicing factor 3a protein complex. The splicing factor 3a heterotrimer includes subunits 1, 2 and 3 and is necessary for the in vitro conversion of 15S U2 snRNP into an active 17S particle that performs pre-mRNA splicing. Subunit 2 interacts with subunit 1 through its amino-terminus while the single zinc finger domain of subunit 2 plays a role in its binding to the 15S U2 snRNP. Subunit 2 may also function independently of its RNA splicing function as a microtubule-binding protein.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: pre-mRNA splicing factor SF3A, subunit 2; SAP 62; SF3a66; spliceosomal protein gene 62; spliceosome associated protein 62; Spliceosome-associated protein 62; Splicing factor 3A subunit 2; splicing factor 3a, subunit 2, 66kD; splicing factor 3a, subunit 2, 66kDa
Gene Aliases: 66kDa; AW047242; PRP11; PRPF11; SAP62; SF3A2; SF3a66; SFA66
UniProt ID: (Human) Q15428, (Rat) Q6AXT8
Entrez Gene ID: (Human) 8175, (Mouse) 20222, (Rat) 299620
Molecular Function:
RNA splicing factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support