Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
SAPCD1 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Protein G7d; Suppressor APC domain-containing protein 1
Gene Aliases: 2810436B06Rik; C6orf26; G7D; NG23; SAPCD1
UniProt ID: (Human) Q5SSQ6, (Mouse) Q9CY86
Entrez Gene ID: (Human) 401251, (Mouse) 78376
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support