Search Thermo Fisher Scientific
FIGURE: 1 / 1
Predicted reactivity based on immunogen sequence: SARS-CoV ORF9b proteins: (50%)
ORF9b plays a role in the inhibition of host innate immune response by targeting the mitochondrial-associated adapter MAVS. Mechanistically, usurps the E3 ligase ITCH to trigger the degradation of MAVS, TRAF3, and TRAF6. In addition, causes mitochondrial elongation by triggering ubiquitination and proteasomal degradation of dynamin-like protein 1/DNM1L.
Protein Aliases: COVID-19 ORF9b; COVID19 ORF9b; ORF9b; SARS-CoV-2 Open Reading Frame 9b; SARS-COV2 ORF9b