Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Aliquoting is unnecessary for -20°C storage.
This gene encodes a member of a highly conserved protein family that exists from archaea to vertebrates and plants. The encoded protein may function in RNA metabolism. Mutations within this gene are associated with Shwachman-Bodian-Diamond syndrome. An alternative transcript has been described, but its biological nature has not been determined. This gene has a closely linked pseudogene that is distally located.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ10917; Protein 22A3; Ribosome maturation protein SBDS; Shwachman-Bodian-Diamond syndrome; Shwachman-Bodian-Diamond syndrome homolog; Shwachman-Bodian-Diamond syndrome protein; Shwachman-Bodian-Diamond syndrome protein homolog
Gene Aliases: 4733401P19Rik; AI836084; CGI-97; SBDS; SDS; SWDS
UniProt ID: (Human) Q9Y3A5, (Mouse) P70122, (Rat) Q5RK30
Entrez Gene ID: (Human) 51119, (Mouse) 66711, (Rat) 288615
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support