Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Aliquoting is unnecessary for -20°C storage.
Mediates the voltage-dependent sodium ion permeability of excitable membranes. Assuming opened or closed conformations in response to the voltage difference across the membrane, the protein forms a sodium-selective channel through which Na+ ions may pass in accordance with their electrochemical gradient. George A.L. Jr., Proc. Natl. Acad. Sci. U.S.A. 89:4893-4897(1992).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Atypical sodium channel Nav2.1; Nax channel; putative voltage-gated sodium channel subunit alpha Nax; Sodium channel Nav2.3; sodium channel protein cardiac and skeletal muscle subunit alpha; Sodium channel protein type 7 subunit alpha; Sodium channel protein type VII subunit alpha; sodium channel, voltage gated, type VII alpha subunit; sodium channel, voltage-gated, type VI, alpha polypeptide; sodium channel, voltage-gated, type VII, alpha subunit; voltage-dependent sodium channel alpha subunit
Gene Aliases: 1110034K09Rik; AI642000; NaG; Nav2; Nav2.1; Nav2.2; Nav2.3; Nax; scl11; SCN6A; SCN7A
UniProt ID: (Human) Q01118, (Mouse) B1AYL1
Entrez Gene ID: (Human) 6332, (Mouse) 20272
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support