Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
23006-1-AP antibody detects the bands around 15 kDa in SDS-PAGE.
Immunogen sequence: MGFPEAASS FRTHQIEAVP TSSASDGFKA NLVFKEIEKK LEEEGEQFVK KIGGIFAFKV KDGPGGKEAT WVVDVKNGKG SVLPNSDKKA DCTITMADSD FLALMTGKMN PQSAFFQGKL KITGNMGLAM KLQNLQLQPG NAKL (405-547 aa encoded by B C005911)
This gene encodes two proteins: sterol carrier protein X (SCPx) and sterol carrier protein 2 (SCP2), as a result of transcription initiation from 2 independently regulated promoters. The transcript initiated from the proximal promoter encodes the longer SCPx protein, and the transcript initiated from the distal promoter encodes the shorter SCP2 protein, with the 2 proteins sharing a common C-terminus. Evidence suggests that the SCPx protein is a peroxisome-associated thiolase that is involved in the oxidation of branched chain fatty acids, while the SCP2 protein is thought to be an intracellular lipid transfer protein. This gene is highly expressed in organs involved in lipid metabolism, and may play a role in Zellweger syndrome, in which cells are deficient in peroxisomes and have impaired bile acid synthesis. Alternative splicing of this gene produces multiple transcript variants, some encoding different isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Acetyl-CoA C-myristoyltransferase; Non-specific lipid-transfer protein; nonspecific lipid transfer protein; NSL-TP; Propanoyl-CoA C-acyltransferase; SCP-2; SCP-2/3-oxoacyl-CoA thiolase; SCP-2/thiolase; SCP-chi; SCP-X; SCPX; Sterol carrier protein 2; Sterol carrier protein X
Gene Aliases: AA409774; AA409893; C76618; C79031; NLTP; ns-LTP; NSL-TP; SCP-2; SCP-CHI; SCP-X; SCP2; SCPX
UniProt ID: (Human) P22307, (Mouse) P32020
Entrez Gene ID: (Human) 6342, (Mouse) 20280
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support