Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LLEDKGLAE VSKVAEQVLN AVNKGLYREA TELWGKAEMI IEQNTDGVNF YNILTKSTPT STMESSLEFT QSHLVCLCQR HVRHLQRDAL SQLMNGPIRK KLKIIPEDQS WGGQATNVFV NMEEDFMKPV ISIVDELLEA GINVTVYNGQ LDLIVDTMGQ EAWVRKLKWP ELPKFSQLKW KALYSDPKSL ETSAFVKSYK NLAFYWILKA GHMVPSDQGD MALKMMRLVT QQE
May be involved in vascular wall and kidney homeostasis. Target protein may act as a serine carboxypeptidase.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: retinoid inducible serine carboxypeptidase; Retinoid-inducible serine carboxypeptidase; Serine carboxypeptidase 1; serine carboxypeptidase 1 precursor protein
Gene Aliases: HSCP1; MSTP034; RISC; SCP1; SCPEP1; UNQ265/PRO302
UniProt ID: (Human) Q9HB40
Entrez Gene ID: (Human) 59342
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support