Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MGSENSALKS YTLREPPFTL PSGLAVYPAV LQDGKFASVF VYKRENEDKV NKAAKHLKTL RHPCLLRFLS CTVEADGIHL VTERVQPLEV ALETLSSAEV CAGIYDILLA LIFLHDRGHL THNNVCLSSV FVSEDGHWKL GGMETVCKVS QATPEFLRSI QSIRDPASIP PEEMSPEFTT LPECHGHARD AFSFGTLVES LLTILNEQVS ADVLSSFQQT LHSTLLNPIP KCRPALCTLL SHDFFRNDFL EVVNFLKSLT LKSEEEKTEF FKFLLDRVSC LSEELIASRL VPLLLNQLVF AEPVAVKSFL PYLLGPKKDH AQGETPCLLS PALFQSRVIP VLLQLFEVHE EHVRMVLLSH IEAYVEHFTQ EQLKKVILPQ VLLGLRDTSD SIVAITLHSL AVLVSLLGPE VVVGGERTKI FKRTAPSFTK NTDLSLEGDP FSQPIKFPIN GLSDVKNTSE DSENFPSSSK KSEEWPDWSE PEEPENQTVN IQIWPREPCD DVKSQCTTLD VEESSWDDCE PSSLDTKVNP GGGITATKPV TSAEQKPIPA LLSLTEESMP WKSSLPQKIS LVQRGDDADQ IEPPKVSSQE RPLKVPSELG LGEEFTIQVK KKPVKDPEMD WFADMIPEIK PSAAFLILPE LRTEMVPKKD DVSPVMQFSS KFAAAEITEG EAEGWEEEGE LNWEDNNW
May play a role in regulating cell adhesion/migration complexes in migrating cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ezrin-binding partner PACE-1 (PACE-1); Ezrin-binding protein PACE-1; Protein-associating with the carboxyl-terminal domain of ezrin; RP1-97P20.2; SCY1-like 3; SCY1-like protein 3; SCY1-like, kinase-like 3
Gene Aliases: PACE-1; PACE1; SCYL3
UniProt ID: (Human) Q8IZE3
Entrez Gene ID: (Human) 57147
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support