Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MDGPVAEHA KQEPFHVVTP LLESWALSQV AGMPVFLKCE NVQPSGSFKI RGIGHFCQEM AKKGCRHLVC SSGGNAGIAA AYAARKLGIP ATIVLPESTS LQVVQRLQGE GAEVQLTGKV WDEANLRAQE LAKRDGWENV PPFDHPLIWK GHASLVQELK AVLRTPPG (1-167 aa encoded by BC009849)
SDSL has low serine dehydratase and threonine dehydratase activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: L-serine deaminase; L-serine dehydratase/L-threonine deaminase; L-threonine dehydratase; SDH; SDH 2; Serine dehydratase 2; serine dehydratase related sequence 1; Serine dehydratase-like; TDH
Gene Aliases: 4432411H13Rik; AI504310; cSDH; SDH 2; SDH1; Sds; SDS-RS1; SDSL; TDH
UniProt ID: (Human) Q96GA7, (Mouse) Q8R238
Entrez Gene ID: (Human) 113675, (Mouse) 257635
Molecular Function:
dehydratase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support