Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 6
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
SEC14L1 may play a role in innate immunity by inhibiting the antiviral RIG-I signaling pathway. In this pathway, functions as a negative regulator of DDX58/RIG-I, the cytoplasmic sensor of viral nucleic acids. Prevents the interaction of DDX58 with MAVS/IPS1, an important step in signal propagation. May also regulate the SLC18A3 and SLC5A7 cholinergic transporters.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: SEC14-like protein 1
Gene Aliases: 1200017E04Rik; 2810012L19Rik; Kiaa4251; Naa-35; PRELID4A; RGD1563123; SEC14L; SEC14L1
UniProt ID: (Human) Q92503, (Mouse) A8Y5H7
Entrez Gene ID: (Human) 6397, (Mouse) 74136, (Rat) 360668
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support