Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSMILSASVI RVRDGLPLSA STDYEQSTGM QECRKYFKML SRKLAQLPDR CTLKTGHYNI NFISSLGVSY MMLCTENYPN VLAFSFLDEL QKEFITTYNM MKTNTAVRPY CFIEFDNFIQ RTKQRYNNPR SLSTKINLSD MQTEIKLRPP YQISMCELGS ANGVTSAFSV DCKGAGKISS AHQRLEPATL SGIVGFILSL LCGALNLIRG FHAIESLLQS DGDDFNYIIA FFLGTAACLY QCYLLVYYTG WRNVKSFLTF GLICLCNMYL YELRNLWQLF FHVTVGAFVT LQIWLRQAQG KAPDYDV
The protein encoded by this gene belongs to the member of the SEC22 family of vesicle trafficking proteins. This protein has similarity to rat SEC22 and may act in the early stages of the secretory pathway. [provided by RefSeq, Nov 2008]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: sec22 homolog; SEC22 vesicle trafficking protein homolog A; SEC22 vesicle trafficking protein-like 2; SEC22 vesicle-trafficking protein homolog A; SEC22 vesicle-trafficking protein-like 2; SEC22A; SEC22L2; Vesicle-trafficking protein SEC22a
Gene Aliases: SEC22A; SEC22L2
UniProt ID: (Human) Q96IW7
Entrez Gene ID: (Human) 26984
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support