Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Sequence of this protein is as follows: MERQEESLSA RPALETEGLR FLHTTVGSLL ATYGWYIVFS CILLYVVFQK LSARLRALRQ RQLDRAAAAV EPDVVVKRQE ALAAARLKMQ EELNAQVEKH KEKLKQLEEE KRRQKIEVWD SMQEGKSYKG NAKKPQEEDS PGPSTSSVLK RKSDRKPLRG GGYNPLSGEG GGACSWRPGR RGPSSGG*G
This gene encodes a transmembrane protein that is localized in the endoplasmic reticulum (ER). It is involved in the degradation process of misfolded proteins in the ER, and may also have a role in inflammation control. It participates in the transfer of misfolded proteins from the ER to the cytosol, where they are destroyed by the proteasome in a ubiquitin-dependent manner. This protein is a selenoprotein, containing the rare amino acid selenocysteine (Sec). Sec is encoded by the UGA codon, which normally signals translation termination. The 3' UTRs of selenoprotein mRNAs contain a conserved stem-loop structure, designated the Sec insertion sequence (SECIS) element, that is necessary for the recognition of UGA as a Sec codon, rather than as a stop signal.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MGC104346; MGC2553; Selenoprotein S; SelS; valosin-containing protein-interacting membrane protein; VCP-interacting membrane protein; VCP-interacting membrane selenoprotein
Gene Aliases: AD-015; ADO15; SBBI8; SELENOS; SELS; SEPS1; VIMP
UniProt ID: (Human) Q9BQE4
Entrez Gene ID: (Human) 55829
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support