Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: CMMVFFLSNM IENQCMSTGA FEITLNDVPV WSKLESGHLP SMQQLVQILD NEMKLNVHMD SIPHHR
Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
SELT encodes a selenoprotein, which contains a selenocysteine (Sec) residue at its active site. The selenocysteine is encoded by the UGA codon that normally signals translation termination. The 3' UTR of selenoprotein genes have a common stem-loop structure, the sec insertion sequence (SECIS), that is necessary for the recognition of UGA as a Sec codon rather than as a stop signal.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: SelT; Thioredoxin reductase-like selenoprotein T
Gene Aliases: SELENOT; SELT; UNQ150/PRO176
UniProt ID: (Human) P62341
Entrez Gene ID: (Human) 51714
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support