Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 6
Recommended positive controls: NT2D1, IMR32, U87-MG, MCF-7, mouse brain.
Predicted reactivity: Mouse (86%), Rat (87%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Inhibits axonal extension by providing local signals to specify territories inaccessible for growing axons. [UniProt]
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Sema C; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 4B; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, 4B; Semaphorin-4B; Semaphorin-C
Gene Aliases: KIAA1745; mKIAA1745; SEMA4B; SEMAC; SemC; UNQ749/PRO1480
UniProt ID: (Human) Q9NPR2, (Mouse) Q62179
Entrez Gene ID: (Human) 10509, (Mouse) 20352
Molecular Function:
membrane-bound signaling molecule
intercellular signal molecule
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support