Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 From June 2-13 take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Sequence of this protein is as follows: RRPSRELDAT PRMTIPYEEL SGTRHFKGQA QNYSTLLLEE ASARLLVGAR GALFSLSAND IGDGAHKEIH WEASPEMQSK CHQKGKNNQT EC
The SEMA4G gene encodes semaphorin 4G, a member of the semaphorin protein family known for their roles in cellular guidance, signaling, and development. Semaphorin 4G is involved in axonal guidance, where it influences the direction and growth of neurons by interacting with receptor complexes such as plexins. This interaction is critical in neural development, contributing to the proper formation of neural circuits by ensuring that neurons connect accurately and functionally. Beyond its role in the nervous system, SEMA4G also participates in immune responses, where it modulates the activity and migration of immune cells, influencing processes like inflammation and immune surveillance. This protein's modulation of cell-to-cell communication and migration extends to its implications in cancer, where altered semaphorin signaling can impact tumor progression and metastasis through changes in cell adhesion and migration properties. Research into SEMA4G continues to investigate its precise mechanisms and interactions, providing insights into therapeutic opportunities in neural disorders and cancer.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: KIAA1619FLJ20590; MGC102867; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 4G; Semaphorin-4G
Gene Aliases: KIAA1619; SEMA4G
UniProt ID: (Human) Q9NTN9
Entrez Gene ID: (Human) 57715
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support