Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: PEDSEPISIS HGNYTKQYPV FVGHKPGRNT TQRHRLDIQM IMIMNGTLYI AARDHIYTVD IDTSHTEEIY CSKKLTWKSR QADVDTCRMK GKHKDECHNF
SEMA6A can act as repulsive axon guidance cues. It may play a role in channeling sympathetic axons into the sympathetic chains and controlling the temporal sequence of sympathetic target innervation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: sema domain, transmembrane domain (TM), and cytoplasmic domain, (semaphorin) 6A; Sema VIA; SEMA6A-1; semaphorin 6A-1; Semaphorin VIA; Semaphorin-6A; Semaphorin-6A-1
Gene Aliases: HT018; KIAA1368; SEMA; SEMA6A; SEMA6A1; SEMAQ; VIA
UniProt ID: (Human) Q9H2E6
Entrez Gene ID: (Human) 57556
Molecular Function: membrane-bound signaling molecule intercellular signal molecule
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support