Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
SEMA6A can act as repulsive axon guidance cues. It may play a role in channeling sympathetic axons into the sympathetic chains and controlling the temporal sequence of sympathetic target innervation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Mutant line 997; sema domain, transmembrane domain (TM), and cytoplasmic domain, (semaphorin) 6A; Sema Q; Sema VIA; SEMA6A-1; semaphorin 6A-1; Semaphorin Q; Semaphorin VIA; Semaphorin-6A; Semaphorin-6A-1
Gene Aliases: 9330158E07; A730020P05Rik; AI851735; b2b997Clo; HT018; KIAA1368; SEMA; SEMA6A; Sema6A-1; SEMA6A1; SEMAQ; VIA
UniProt ID: (Human) Q9H2E6, (Mouse) O35464
Entrez Gene ID: (Human) 57556, (Rat) 361324, (Mouse) 20358
Molecular Function:
membrane-bound signaling molecule
intercellular signal molecule
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support