Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: RDTRVYGVF SNPWNYSAVC VYSLGDIDKV FRTSSLKGYH SSLPNPRPGK CLPDQQPIPT ETFQVADRHP EVAQRVEPMG PLKTPLFHSK YHYQKVAVHR MQASHGETFH VLYLTTDRGT IHKVVEPGEQ EHSFAFNIME IQPFRRAAAI QTMSLDAERR KLYVSSQWEV SQVPLDLCEV YGGGCHGCLM SRDPYCGWDQ GRCISIYSSE RSVLQSINPA EPHKECPNPK PDKAPLQKVS LAPNSRYYLS CPMESRHATY SWRHKENVEQ SCEPGHQSPN CILFIENLTA QQYGHYFCEA QEGSYFREAQ HWQLLPEDGI MAEHLLGHAC ALAASLWLGV LPTLTLGLLV H (317-666 aa encoded by BC101643)
CD108 (Sema7A) is a GPI-anchored semaphorin family member, which enhances central and peripheral axonal growth and is required for proper axon track formation during ebryogenesis. CD108 also regulates osteoclast differentiation and pre-osteoblastic cell migration, and in immune system affects cell proliferation, chemotaxis and cytokine release. On erythrocytes CD108 defines the JMH (John-Milton-Hagen) human blood group. CD108 signalizes through its receptors - plexin C1 and beta1 integrins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD108; CDw108; H-Sema K1; JMH blood group antigen; John Milton Hagen blood group H-Sema K1; John-Milton-Hargen human blood group Ag; MGC126692; MGC126696; sema domain, immunoglobulin domain (Ig), and GPI membrane anchor, (semaphorin) 7A (JMH blood group); sema domain, immunoglobulin domain (Ig), and GPI membrane anchor, 7A; Sema K1; Sema L; semaphorin 7A, GPI membrane anchor (John Milton Hagen blood group); Semaphorin K1; Semaphorin-7A; Semaphorin-K1; Semaphorin-L
Gene Aliases: 2900057C09Rik; CD108; CDw108; H-SEMA-K1; H-Sema-L; JMH; M-Sema-L; SEMA7A; SEMAK1; SEMAL; Semk1
UniProt ID: (Human) O75326, (Mouse) Q9QUR8
Entrez Gene ID: (Human) 8482, (Mouse) 20361
Molecular Function:
membrane-bound signaling molecule
intercellular signal molecule
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support