Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Positive test controls include: rat lung, rat testis.
Immunogen sequence: LELTEDMEKE ISNALGHGPQ DEILSSAFKL RITRGDIQTL KNYHWLNDEV INFYMNLLVE RNKKQGYPAL HVFSTFFYPK LKSGGYQAVK RWTKGVNLFE QEIILVPIHR KVHWSLVVID LRKKCLKYLD SMGQKGHRIC EILLQYLQDE SKTKRNSDLN LLEWTHHSMK PHEIPQQLNG SDCGMFTCKY ADYISRDKPI TFTQHQMPLF RKKMVWEILH QQLL
SUMO is a small ubiquitin-like protein that can be covalently conjugated to other proteins. SENP2 is one of a group of enzymes that process newly synthesized SUMO1, SUMO2, and SUMO3 into the conjugatable mature forms and catalyze the deconjugation of these same SUMO proteins from their targeted substrates. SENP2 may also down-regulate CTNNB1 levels and thereby modulate the Wnt pathway.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Axam; Axam2; Axin-associating molecule; CT; Sentrin-specific protease 2; Sentrin/SUMO-specific protease SENP2; Smt3ip2; SUMO-1 protease 1; SUMO-1 protease-1; SUMO-1/Smt3-specific isopeptidase 2; SUMO/sentrin specific peptidase 2; SUMO/sentrin specific protease 2; SuPr-1
Gene Aliases: 2310007L05Rik; 4930538C18Rik; AI646780; AW554757; Axam; mKIAA1331; Senp2; Smt3ip2; SuPr-1; Supr1
UniProt ID: (Mouse) Q91ZX6, (Rat) Q9EQE1
Entrez Gene ID: (Mouse) 75826, (Rat) 78973
Molecular Function:
protease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support