Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This gene encodes a member of the Jumonji, AT-rich interactive domain 1 (JARID1) histone demethylase protein family. The encoded protein plays a role in gene regulation through the histone code by specifically demethylating lysine 4 of histone H3. The encoded protein interacts with many other proteins, including retinoblastoma protein, and is implicated in the transcriptional regulation of Hox genes and cytokines. This gene may play a role in tumor progression.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 7-Sep; CDC10 (cell division cycle 10, S. cerevisiae, homolog); CDC10 (cell division cycle 10, S.cerevisiae, homolog); CDC10 protein homolog; cell division cycle 10 homolog; septin 7 variant 4; Septin-7
Gene Aliases: CDC10; CDC3; E430034N22; NBLA02942; SEPT7; SEPT7A; SEPTIN7
UniProt ID: (Human) Q16181, (Mouse) O55131, (Rat) Q9WVC0
Entrez Gene ID: (Human) 989, (Mouse) 235072, (Rat) 64551
Molecular Function:
cytoskeletal protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support