Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 2
Immunogen sequence: HFLILYVEPL PLIFQITPLN VTQWLMVLKI SLPVILMDET LKFVARNYLE PGKECVQPAT KSCSFSACTD GISWPFVLLI MPLVIWVYST DTNFSDMFWS
This gene encodes one of the SERCA Ca(2+)-ATPases, which are intracellular pumps located in the sarcoplasmic or endoplasmic reticula of muscle cells. This enzyme catalyzes the hydrolysis of ATP coupled with the translocation of calcium from the cytosol into the sarcoplasmic reticulum lumen, and is involved in regulation of the contraction/relaxation cycle. Mutations in this gene cause Darier-White disease, also known as keratosis follicularis, an autosomal dominant skin disorder characterized by loss of adhesion between epidermal cells and abnormal keratinization. Alternative splicing results in multiple transcript variants encoding different isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATPase Ca++ transporting cardiac muscle slow twitch 2; ATPase, Ca++ dependent, slow-twitch, cardiac muscle-2; ATPase, Ca++ transporting, cardiac muscle, slow twitch 2; ATPase, Ca++ transporting, slow twitch 2; Calcium pump 2; Calcium-transporting ATPase sarcoplasmic reticulum type, slow twitch skeletal muscle isoform; cardiac Ca2+ ATPase; Endoplasmic reticulum class 1/2 Ca(2+) ATPase; sarco(endo)plasmic reticulum Ca(2+)-dependent ATPase 2; sarco/endoplasmic reticulum Ca2+-ATPase 2; Sarcoplasmic/endoplasmic reticulum calcium ATPase 2; SERCA; SERCA ATPase; SERCA2; SR Ca(2+)-ATPase 2
Gene Aliases: 9530097L16Rik; ATP2A2; ATP2B; D5Wsu150e; DAR; DD; mKIAA4195; SERCA2; Serca2a; SERCA2B; SercaII
UniProt ID: (Human) P16615, (Mouse) O55143, (Rat) P11507
Entrez Gene ID: (Human) 488, (Mouse) 11938, (Rat) 29693
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support