Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide Sequence: KHGQLANEAA FLPVPQKIEA HCFQNEKVTA IWSGWTHLVA QTETGKMFTW GRADYGQLGR KLETYEGWKL EKQDSFLPCS RPPNSMPSSP HCLTGATE
Probable guanine nucleotide exchange factor (GEF), which may be involved in the secretion process.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: deafness locus associated putative guanine nucleotide exchange factor; Deafness locus-associated putative guanine nucleotide exchange factor; DelGEF; Guanine nucleotide exchange factor-related protein; Secretion-regulating guanine nucleotide exchange factor
Gene Aliases: DELGEF; GNEFR; SERGEF
UniProt ID: (Human) Q9UGK8
Entrez Gene ID: (Human) 26297
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support