Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Sequence of this protein is as follows: MKVVPSLLLS VLLAQVWLVP GLAPSPQSPE TPAPQNQTSR VVQAPKEEEE DEQEASEEKA SEEEKAWLMA SRQQLAKETS NFGFSLLRKI SMRHDGNMVF SPFGMSLAMT GLMLGATGPT ETQIKRGLHL QALKPTKPGL LPSLFKGLRE TLSRNLELGL TQGSFAFIHK DFDVKETFFN LSKRYFDTEC VPMNFRNASQ AKRLMNHYIN KETRGKIPKL FDEINPETKL ILVDYILFKG KWLTPFDPVF TEVDTFHLDK YKTIKVPMMY GAGKFASTFD KNFRCHVLKL PYQGNATMLV VLMEKMGDHL ALEDYLTTDL VETWLRNMKT RNMEVFFPKF KLDQKYEMHE LLRQMGIRRI FSPFADLSEL SATGRNLQVS RVLQRTVIEV DERGTEAVAG ILSEITAYSM PPVIKVDRPF HFMIYEETSG MLLFLGRVVN PTLL
The protein encoded by this gene belongs to the serpin family. It is predominantly expressed in the liver and secreted in plasma. It inhibits the activity of coagulation factors Xa and XIa in the presence of protein Z, calcium and phospholipid. Mutations in this gene are associated with venous thrombosis. Alternatively spliced transcript variants have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Protein Z-dependent protease inhibitor; PZ-dependent protease inhibitor; serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 10; Serpin A10; serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 10
Gene Aliases: PZI; SERPINA10; UNQ707/PRO1358; ZPI
UniProt ID: (Human) Q9UK55
Entrez Gene ID: (Human) 51156
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support