Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
This gene encodes a member of the serine proteinase inhibitor (serpin) superfamily. This member is the principal inhibitor of tissue plasminogen activator (tPA) and urokinase (uPA), and hence is an inhibitor of fibrinolysis. Defects in this gene are the cause of plasminogen activator inhibitor-1 deficiency (PAI-1 deficiency), and high concentrations of the gene product are associated with thrombophilia. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Acrosomal serine protease inhibitor; antitr; PAI-3; PCI; Plasma serine protease inhibitor; Plasminogen activator inhibitor 3; plasminogen activator inhibitor III; plasminogen activator inhibitor-3; Protein C inhibitor; serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 5; Serpin A5; serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 5
Gene Aliases: PAI-3; PAI3; PCI; PCI-B; PLANH3; PROCI; SERPINA5
UniProt ID: (Human) P05154
Entrez Gene ID: (Human) 5104
Molecular Function: protease inhibitor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support