Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Sequence of this protein is as follows: MPLLLYTCLL WLPTSGLWTV QAMDPNAAYV NMSNHHRGLA SANVDFAFSL YKHLVALSPK KNIFISPVSI SMALAMLSLG TCGHTRAQLL QGLGFNLTER SETEIHQGFQ HLHQLFAKSD TSLEMTMGNA LFLDGSLELL ESFSADIKHY YESEVLAMNF QDWATASRQI NSYVKNKTQG KIVDLFSGLD SPAILVLVNY IFFKGTWTQP FDLASTREEN FYVDETTVVK VPMMLQSSTI SYLHDAELPC QLVQMNYVGN GTVFFILPDK GKMNTVIAAL SRDTINRWSA GLTSSQVDLY IPKVTISGVY DLGDVLEEMG IADLFTNQAN FSRITQDAQL KSSKVVHKAV LQLNEEGVDT AGSTGVTLNL TSKPIILRFN QPFIIMIFDH FTWSSLFLAR VMNPV
This gene encodes an alpha-globulin protein with corticosteroid-binding properties. This is the major transport protein for glucorticoids and progestins in the blood of most vertebrates. The gene localizes to a chromosomal region containing several closely related serine protease inhibitors which may have evolved by duplication events.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CBG; corticosteroid binding globulin; Corticosteroid-binding globulin; serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 6; Serpin A6; serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 6; Transcortin
Gene Aliases: CBG; SERPINA6
UniProt ID: (Human) P08185
Entrez Gene ID: (Human) 866
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support