Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Members of the human serpin family regulate a diverse array of serine and cysteine proteinases associated with essential biological processes such as fibrinolysis, coagulation, inflammation, cell mobility, cellular differentiation, and apoptosis. Most serpins are secreted and attain physiologic concentrations in the blood and extracellular fluids. However, a subset of the serpin superfamily, the ov-serpins, also resides intracellularly. Differential display was used to identify a novel serpin (headpin or SERPINB13) that is under expressed in squamous cell cancers of the oral cavity. SERPINB13 is expressed in normal oral mucosal tissue, skin, and cultured keratinocytes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HaCaT UV-repressible serpin; Headpin; HURPIN; Peptidase inhibitor 13; PI-13; protease inhibitor 13 (hurpin, headpin); Proteinase inhibitor 13; serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 13; Serpin B13; serpin peptidase inhibitor, clade B (ovalbumin), member 13; UV-B repressed sequence, HUR 7
Gene Aliases: headpin; HSHUR7SEQ; HUR7; PI13; SERPINB13
UniProt ID: (Human) Q9UIV8
Entrez Gene ID: (Human) 5275
Molecular Function:
protease inhibitor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support