Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
MA5-17648 targets PI-9/Proteinase Inhibitor 9 in ELISA, FACS, and WB applications and shows reactivity with Human samples.
The MA5-17648 immunogen is recombinant PI-9 produced in P. pastoris.
PI9 belongs to the large superfamily of serine proteinase inhibitors (serpins), which bind to and inactivate serine proteinases. These interactions are involved in many cellular processes, including coagulation, fibrinolysis, complement fixation, matrix remodeling, and apoptosis (Sprecher et al., 1995 [PubMed 8530382]).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CAP-3; Cytoplasmic antiproteinase 3; Peptidase inhibitor 9; PI-9; protease inhibitor 9 (ovalbumin type); serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 9; Serpin B9; serpin peptidase inhibitor, clade B (ovalbumin), member 9; serpin peptidase inhibitor, clade B, member 9; testicular tissue protein Li 180
Gene Aliases: CAP-3; CAP3; PI-9; PI9; SERPINB9
UniProt ID: (Human) P50453
Entrez Gene ID: (Human) 5272
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support