Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: EELGSNTGI QVFNQIVKSR PHDNIVISPH GIASVLGMLQ LGADGRTKKQ LAMVMRYGVN GVGKILKKIN KAIVSKKNKD IVTVANAVFV KNASEIEVPF VTRNKDVFQC EVRNVNFEDP ASACDSINAW VKNETRDMID NLLSPDLIDG VLTRLVLVNA VYFKGLWKSR FQPENTKKRT FVAADGKSYQ VPMLAQLSVF RCGSTSAPND LWYNFIELPY HGESISMLIA LPTESSTPLS AIIPHISTKT IDSWMSIMVP KRVQVILPKF TAVAQTDLKE PLKVLGITDM FDSSKANFAK ITTGSENLHV SHILQKAKIE VSEDGTKASA ATTAILIARS SPPWFIVDRP FLFFIRHNPT GAVLFMGQIN K (28-397 aa encoded by BC015663)
This gene encodes a member of the serpin family of proteins, a group of proteins that inhibit serine proteases. Thrombin, urokinase, plasmin and trypsin are among the proteases that this family member can inhibit. This gene is a susceptibility gene for chronic obstructive pulmonary disease and for emphysema. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GDN; Glia derived nexin; Glia-derived nexin; Glia-derived nexin/protease nexin I; glial-derived neurite promoting factor; P17; Peptidase inhibitor 7; PI-7; plasminogen activator inhibitor type 1; PN-1; Protease nexin 1; Protease nexin I; protease nexin PN-1; serine (or cysteine) peptidase inhibitor, clade E, member 2; serine (or cysteine) proteinase inhibitor, clade E (nexin, plasminogen activator inhibitor type 1), member 2; serine (or cysteine) proteinase inhibitor, clade E, member 2; serine protease inhibitor 4; Serine protease-inhibitor 4; Serpin E2; serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitor type 1), member 2; serpin peptidase inhibitor, clade E, member 2; SERPINE 2
Gene Aliases: B230326M24Rik; GDN; GDNPF; Gdnpn1; PAI-1; PI-7; PI7; PN-1; PN1; PNI; SERPINE2; Spi4; Spin4
UniProt ID: (Human) P07093, (Rat) P07092, (Mouse) Q07235
Entrez Gene ID: (Human) 5270, (Rat) 29366, (Mouse) 20720
Molecular Function: protease inhibitor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support