Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Immunogen sequence: VPTHFEWNVS QVLANLSWDT LHPPLVWERP TKVRLPKLYL KHQMDLVATL SQLGLQELFQ APDLRGISEQ SLVVSGVQHQ STLELSEVGV EAAAATSIAM SRMSLSSFSV NRPFLFFIFE DTTGLPLFVG SVRNPNPSAP RELKEQQDSP GNKDFLQSLK GFPRGDKLFG PDLKLVPPME EDYPQFGSPK; Positive Samples: HepG2, A-549, Rat liver
This gene encodes a member of the serpin family of serine protease inhibitors. The protein is a major inhibitor of plasmin, which degrades fibrin and various other proteins. Consequently, the proper function of this gene has a major role in regulating the blood clotting pathway. Mutations in this gene result in alpha-2-plasmin inhibitor deficiency, which is characterized by severe hemorrhagic diathesis. Multiple transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: alpha 2 antiplasmin; Alpha-2-antiplasmin; Alpha-2-AP; Alpha-2-PI; Alpha-2-plasmin inhibitor; pigment epithelium derived factor; plasmin inhibitor alpha 2; serine (or cysteine) peptidase inhibitor, clade F, member 2; serine (or cysteine) proteinase inhibitor, clade F, member 2; Serpin F2
Gene Aliases: AI747498; Pli; Serpimf2; Serpinf2
UniProt ID: (Mouse) Q61247
Entrez Gene ID: (Mouse) 18816, (Rat) 287527
Molecular Function:
protease inhibitor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support