Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MRSLLLLSAF CLLEAALAAE VKKPAAAAAP GTAEKLSPKA ATLAERSAGL AFSLYQAMAK DQAVENILVS PVVVASSLGL VSLGGKATTA SQAKAVLSAE QLRDEEVHAG LGELLRSLSN STARNVTWKL GSRLYGPSSV SFADDFVRSS KQHYNCEHSK INFRDKRSAL QSINEWAAQT TDGKLPEVTK DVERTDGALL VNAMFFKPHW DEKFHHKMVD NRGFMVTRSY TVGVMMMHRT GLYNYYDDEK EKLQIVEMPL AHKLSSLIIL MPHHVEPLER LEKLLTKEQL KIWMGKMQKK AVAISLPKGV VEVTHDLQKH LAGLGLTEAI DKNKADLSRM SGKKDLYLAS VFHATAFELD TDGNPFDQDI YGREELRSPK LFYADHPFIF LVRDTQSGSL LFIGRLVRPK GDKMRDEL
SERPINH1 is a member of the serpin superfamily of serine proteinase inhibitors, and its expression is induced by heat shock. The SERPINH1 protein localizes to the endoplasmic reticulum lumen and binds collagen; thus it is thought to be a molecular chaperone involved in the maturation of collagen molecules. Autoantibodies to SERPINH1 have been found in patients with rheumatoid arthritis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 47 kDa heat shock protein; Arsenic-transactivated protein 3; AsTP 3; AsTP3; CBP 1; CBP 2; Cell proliferation-inducing gene 14 protein; collagen binding protein 1; collagen binding protein 2; Collagen-binding protein; Colligen; Colligin; Colligin 1; Colligin 2; colligin-1; colligin-2; GP46; HGNC 1547; Hsp 47; PIG 14; RA A47; rheumatoid arthritis antigen A-47; Rheumatoid arthritis-related antigen RA-A47; serine (or cysteine) peptidase inhibitor, clade H, member 1; serine (or cysteine) proteinase inhibitor, clade H (heat shock protein 47), member 1; serine (or cysteine) proteinase inhibitor, clade H (heat shock protein 47), member 1, (collagen binding protein 1); serine (or cysteine) proteinase inhibitor, clade H (heat shock protein 47), member 2; serine (or cysteine) proteinase inhibitor, clade H (heat shock protein 47), member 2, (collagen-binding protein 2); serine (or cysteine) proteinase inhibitor, clade H, member 1; serine proteinase inhibitor, clade H (heat shock protein 47), member 1; Serpin H1; serpin peptidase inhibitor, clade H (heat shock protein 47), member 1, (collagen binding protein 1); serpin peptidase inhibitor, clade H, member 1
Gene Aliases: AsTP3; CBP1; CBP2; gp46; HSP47; OI10; PIG14; PPROM; RA-A47; SERPINH1; SERPINH2
UniProt ID: (Human) P50454, (Rat) P29457
Entrez Gene ID: (Human) 871, (Rat) 29345
Molecular Function:
protease inhibitor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support