Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MNRGGGSPSA AANYLLCTNC RKVLRKDKRI RVSQPLTRGP SAFIPEKEVV QANTVDERTN FLVEEYSTSG RLDNITQVMS LHTQYLESFL RSQFYMLRMD GPLPLPYRHY IAIMAAARHQ CSYLINMHVD EFLKTGGIAE WLNGLEYVPQ RLKNLNEINK LLAHRPWLIT KEHIQKLVKT GENNWSLPEL VHAVVLLAHY HALASFVFGS GINPERDPEI SNGFRLISVN NFCVCDLAND NNIENASLSG SNFGIVDSLS ELEALMERMK RLQEEREDEE ASQEEMSTRF EKEKKESLFV VSGDTFHSFP HSGAFLHFFA F
May function as an intracellular leucine sensor that negatively regulates the TORC1 signaling pathway (PubMed:25263562). May also regulate the insulin-receptor signaling pathway through activation of TORC2. This metabolic regulator may also play a role in protection against oxidative and genotoxic stresses.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Sestrin-3
Gene Aliases: SESN3; SEST3
UniProt ID: (Human) P58005
Entrez Gene ID: (Human) 143686
Molecular Function: peroxidase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support