Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: KRLTLETMK MMLDKKQIRA IFLFEFKMGR KAAETTRNIN NAFGPGTANE RTVQWWFKKF CKGDESLEDE ERSGRPSEVD NDQLRAIIEA DPLTTTREVA EELNVNHSTV VRHLKQIGKV KKLDKWVPHE LTENQKNRRF EVSSSLILRN HNEPFLDRIV TCDEKWILYD NRRRSAQWLD QEEAPKHFPK PILHPKKVMV TIWWSAAGLI HYSFLNPGET ITSEKYAQEI DEMNQKLQRL QLALVNRKGP ILLHDNARPH VAQPTLQKLN ELGYEVLPHP PYSPDLLPTN YHVFKHLNNF LQGKRFHNQQ DAENAFQEFV ESQSTDFYAT GINQLISRWQ KCVDCNGSYF D (322-671 aa encoded by BC008931)
SETMAR (SET domain and mariner transposase fusion gene) is a histone methyltransferase that methylates Lys-4 and Lys-36 of histone H3, creating specific tags for epigenetic transcriptional activation. It specifically mediates dimethylation of H3 Lys-36. SETMAR has sequence-specific DNA-binding activity, and recognizes the 19-mer core of the 5'-terminal inverted repeats (TIRs) of the Hsmar1 transposable element. SETMAT has DNA nicking activity, and in vivo end-joining activity, and may mediate genomic integration of foreign DNA.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Histone-lysine N-methyltransferase SETMAR; Metnase; SET domain and mariner transposase fusion gene-containing protein; SET domain and mariner transposase fusion protein
Gene Aliases: Mar1; METNASE; SETMAR
UniProt ID: (Human) Q53H47
Entrez Gene ID: (Human) 6419
Molecular Function: viral or transposable element protein histone modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support