Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAEEMVEAAG EDERELAAEM AAAFLNENLP ESIFGAPKAG NGQWASVIRV MNPIQGNTLD LVQLEQNEAA FSVAVCRFSN TGEDWYVLVG VAKDLILNPR SVAGGFVYTY K
This gene encodes subunit 3 of the splicing factor 3b protein complex. Splicing factor 3b, together with splicing factor 3a and a 12S RNA unit, forms the U2 small nuclear ribonucleoproteins complex transcription coactivator-HAT complex, and the TFTC -containing complex). These complexes may function in chromatin modification, transcription, splicing, and DNA repair.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: pre-mRNA splicing factor SF3b, 130 kDa subunit; Pre-mRNA-splicing factor SF3b 130 kDa subunit; SAP 130; SF3b130; Spliceosome-associated protein 130; Splicing factor 3B subunit 3; STAF130
Gene Aliases: KIAA0017; RSE1; SAP130; SF3b130; SF3B3; STAF130
UniProt ID: (Human) Q15393
Entrez Gene ID: (Human) 23450
Molecular Function:
RNA processing factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support