Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: MTDRYTIHS QLEHLQSKYI GTGHADTTKW EWLVNQHRDS YCSYMGHFDL LNYFAIAENE SKARVRFNLM EKMLQPCGPP ADKPEEN (1-86 aa encoded by B C000198)
SF3B5 is a protein coding gene involved in pre-mRNA splicing as a component of the splicing factor SF3B complex, a constituent of the spliceosome. SF3B complex is required for 'A' complex assembly formed by the stable binding of U2 snRNP to the branchpoint sequence (BPS) in pre-mRNA. Sequence independent binding of SF3A/SF3B complex upstream of the branch site is essential, it may anchor U2 snRNP to the pre-mRNA.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MGC3133; Pre-mRNA-splicing factor SF3b 10 kDa subunit; SF3b5; Splicing factor 3B subunit 5; splicing factor 3b, subunit 5, 10kDa
Gene Aliases: 10kDa; 1110005L13Rik; AU043053; SF3B10; SF3B5; Ysf3
UniProt ID: (Human) Q9BWJ5, (Mouse) Q923D4
Entrez Gene ID: (Human) 83443, (Mouse) 66125, (Rat) 680891
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support