Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: SSRKQPMSAT LRERLRKTRF SFNSSYNVVK RLKVESEEND QTFSEKPASS TEENCLEFQE SFKHIDSEFE ENTNLKNTLK NLNVCESQ
Highest antigen sequence identity to the following orthologs: Mouse - 50%, Rat - 52%.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MEI5 recombination repair protein homolog; Meiosis protein 5 homolog; SWI5-dependent homologous recombination repair protein 1; Swi5-dependent recombination DNA repair protein 1 homolog; SWI5-dependent recombination repair 1; SWI5-dependent recombination repair protein 1
Gene Aliases: bA373N18.1; C10orf78; MEI5; MEIR5; SFR1
UniProt ID: (Human) Q86XK3
Entrez Gene ID: (Human) 119392
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support