Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAAATIVHD TSEAVELCPA YGLYLKPITK MTISVALPQL KQPGKSISNW EVMERLKGMV QNHQFSTLRI SKSTMDFIRF EGEVENKSLV KSFLACLDGK TIKLSGFSDI LKVRAAEFKI DFPTRHDWDS FFRDAKDMNE TLPGERPDTI HLEGLPCKWF ALKESGSEKP SEDVLVKVFE KFGEIRNVDI PMLDSYREEM TGRNFHTFSF GGHLNFEAYV QYREYMGFIQ AMSALRGMKL MYKGEDGKAV ACNIKVSFDS TKHLSDASIK KRQLERQKLQ ELEQQREEQK RREKEAEERQ RAEERKQKEL EELERERKRE EKLRKREQKQ RDRELRRNQK KLEKLQAEEQ K (1-350 aa encoded by BC028151)
DXYS155E is a gene found in the pseudoautosomal region of the distal short arms of the X and Y chromosomes, and appears to be ubiquitously expressed.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 721P; A kinase (PRKA) anchor protein 17A; A-kinase anchor protein 17A; AKAP-17A; B-lymphocyte antigen; B-lymphocyte surface antigen; PRKA17A; Protein kinase A-anchoring protein 17A; Protein XE7; pseudoautosomal gene XE7; RP13-297E16.1; Splicing factor, arginine/serine-rich 17A
Gene Aliases: 721P; AKAP-17A; AKAP17A; CCDC133; CXYorf3; DXYS155E; PRKA17A; SFRS17A; XE7; XE7Y
UniProt ID: (Human) Q02040
Entrez Gene ID: (Human) 8227
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support