Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody detects 130-150 kDa SFRS8 protein.
Immunogen sequence: LDDDSDDDE ESKEGQESSS SAANTNPAVA PPCVVVEEKK PQLTQEELEA KQAKQKLEDR LAAAAREKLA QASKESKEKQ LQAERKRKAA LFLQTLKNPL PEAEAGKIEE SPFSVEESST TPCPLLTGGR PLPTLEVKPP DRPSSKSKDP PREEEKEKKK KKHKKRSRTR SRSPKYHSSS KSRSRSHSKA KHSLPSAYRT VRRSRSRSRS PRRRAHSPER RREERSVPTA YRVSRSPGAS RKRTRSRSPH EKKKKRRSRS RTKSKARSQS VSPSKQAAPR PAAPAAHSAH SASVSPVESR GSSQERSRGV SQEKEAQISS AIVSSVQSKI TQDLMAKVRA MLAASKNLQT SAS (600-951 aa encoded by BC136678)
Signal-induced proliferation associated-like protein 2 (SIPA1L2) is a member of the SIPA1 family of RapGAPs. Little is known of the role of the SIPA1L2 protein, but recent studies of SIPA indicate that its deregulation can cause myeloproliferative stem cell disorders in mice and increased metastases in human cancers. Other studies suggest SIPA1L1 may play important roles in embryo development and control of cell proliferation. Based on the amount of homology between SIPA family members, it is likely that SIPA1L2 plays a role in embryo development and cell proliferation, possibly including oncogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Splicing factor, arginine/serine-rich 8; splicing factor, arginine/serine-rich 8 (suppressor-of-white-apricot homolog, Drosophila); splicing factor, arginine/serine-rich 8 (suppressor-of-white-apricot, Drosophila homolog); splicing factor, suppressor of white-apricot family; Splicing factor, suppressor of white-apricot homolog; Suppressor of white apricot protein homolog
Gene Aliases: SFRS8; SFSWAP; SWAP
UniProt ID: (Human) Q12872
Entrez Gene ID: (Human) 6433
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support