Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 µg/mL.
Surfactant protein D (SP-D) is synthesized and secreted by lung epithelial cells. It belongs to group III of the family of C-type lectins and members of this group has overall structure consisting of multiple globular 'head' regions linked by triple-helical, collagen-like, strands. This group also includes SP-A and the serum proteins mannan-binding protein, conglutinin and collectin-43, all of which have been shown to bind to the C1q receptor found on a wide variety of cells. Both SP-D and SP-A have been shown to enhance oxygen radical production by alveolar macrophages.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Collectin-7; Collectin7; CP4; Lung surfactant protein D; PSP-D; PSP-D Surfactant protein D; pulmonary surfactant apoprotein; Pulmonary surfactant associated protein PSP-D; pulmonary surfactant protein D; Pulmonary surfactant-associated protein D; SFTP 4; surfactant associated protein D; Surfactant associated protein pulmonary 4; surfactant, pulmonary-associated protein D; surfactant-associated protein, pulmonary 4
Gene Aliases: COLEC7; PSP-D; PSPD; SFTP4; SFTPD; SP-D; SPD
UniProt ID: (Human) P35247, (Rat) P35248
Entrez Gene ID: (Human) 6441, (Rat) 25350
Molecular Function: surfactant
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support