Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: EEGRAVHGA APQDLSRNLE FSELPDHCYR MNSSPAGTPS PQPSRANGNI NLGPSANPNA QPTDFDFLKV IGKGNYGKVL LAKRKSDGAF YAVKVLQKKS ILKKKEQSHI MAERSVLLKN VRHPFLVGLR YSFQTPEKLY FVLDYVNGGE LFFHLQRERR FLEPRARFYA AEVASAIGYL HSLNIIYRDL KPENILLDCQ GHVVLTDFGL CKEGVEPEDT TSTFCGTPEY LAPEVLRKEP YDRAVDWWCL GAVLYEMLHG LPPFYSQDVS QMYENILHQP LQIPGGRTVA ACDLLQSLLH KDQRQRLGSK ADFLEIKNHV FFSPINWDDL YHKRLTPPFN PNVTGPADLK HFDPEFTQEA VSKSIGCTPD TVASSSGASS AFLGFSYAPE DDDILDC (1-397 aa encoded by BC014037)
SGK2 is a serine/threonine protein kinase which is similar to serum- and glucocorticoid-induced protein kinase (SGK). Unlike SGK, SGK2 is not induced by serum or glucocorticoids. It is induced in response to signals that activate phosphatidylinositol 3-kinase, which is also true for SGK. Although little is known about the transcriptional regulation of SGK2, the protein has been shown to be phosphorylated and activated by 3 phosphoinositide dependent protein kinase 1 (PDK1), and to a lesser extent insulin like growth factor 1, in vitro.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Serine/threonine-protein kinase Sgk2; serum/glucocorticoid regulated kinase 2; Serum/glucocorticoid-regulated kinase 2
Gene Aliases: AI098171; AW146006; dJ138B7.2; H-SGK2; SGK2; Sgkl
UniProt ID: (Human) Q9HBY8, (Rat) Q8R4U9, (Mouse) Q9QZS5
Entrez Gene ID: (Human) 10110, (Rat) 171497, (Mouse) 27219
Molecular Function:
non-receptor serine/threonine protein kinase
protein modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support