Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • SGK3 Antibodies

            Proteintech

            SGK3 Polyclonal Antibody

            2 References
            View all (13) SGK3 antibodies
            Datasheet
            Protocols
            Questions & Answers
            Datasheet
            Protocols
            Questions & Answers

            Cite SGK3 Polyclonal Antibody

            • Antibody Testing Data (10)
            Application
            SGK3 Antibody in Immunocytochemistry (ICC/IF)
            Group 53 Created with Sketch.

            FIGURE: 1 / 10

            SGK3 Antibody (12699-1-AP) in ICC/IF

            Immunofluorescent analysis of A375 cells using 12699-1-AP (SGK3 antibody) at dilution of 1:25 and Alexa Fluor 488-conjugated AffiniPure Goat Anti-Rabbit IGG (H+L).
            View Product

            SGK3 Antibody in Immunocytochemistry (ICC/IF)
            SGK3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            SGK3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            SGK3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            SGK3 Antibody in Western Blot (WB)
            SGK3 Antibody in Western Blot (WB)
            SGK3 Antibody in Western Blot (WB)
            SGK3 Antibody in Western Blot (WB)
            SGK3 Antibody in Western Blot (WB)
            SGK3 Antibody in Immunoprecipitation (IP)

            Product Details

            12699-1-AP

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            1:500-1:2,000
            View 1 publication 1 publication

            Immunohistochemistry (Paraffin) (IHC (P))

            1:20-1:200
            -

            Immunocytochemistry (ICC/IF)

            1:10-1:100
            View 1 publication 1 publication

            Immunoprecipitation (IP)

            0.5-4.0 µg
            -
            Product Specifications

            Species Reactivity

            Human, Mouse, Rat

            Host/Isotype

            Rabbit / IgG

            Class

            Polyclonal

            Type

            Antibody

            Immunogen

            SGK3 Fusion Protein Ag3149 (160-496 aa encoded by BC015326)
            View immunogen

            Conjugate

            Unconjugated

            Form

            Liquid

            Concentration

            0.25 mg/mL

            Purification

            Antigen affinity chromatography

            Storage buffer

            PBS, pH 7.3, with 50% glycerol

            Contains

            0.02% sodium azide

            Storage conditions

            -20°C

            Shipping conditions

            Wet ice (domestic); Dry ice (international)

            Product Specific Information

            Immunogen sequence: TDFDFLKVI GKGSFGKVLL AKRKLDGKFY AVKVLQKKIV LNRKEQKHIM AERNVLLKNV KHPFLVGLHY SFQTTEKLYF VLDFVNGGEL FFHLQRERSF PEHRARFYAA EIASALGYLH SIKIVYRDLK PENILLDSVG HVVLTDFGLC KEGIAISDTT TTFCGTPEYL APEVIRKQPY DNTVDWWCLG AVLYEMLYGL PPFYCRDVAE MYDNILHKPL SLRPGVSLTA WSILEELLEK DRQNRLGAKE DFLEIQNHPF FESLSWADLV QKKIPPPFNP NVAGPDDIRN FDTAFTEETV PYSVCVSSDY SIVNASVLEA DDAFVGFSYA PPSEDLFL (160-496 aa encoded by BC015326)

            Target Information

            SGKL (SGK3) is a serine/threonine protein kinase which is similar to serum- and glucocorticoid-induced protein kinase (SGK). Unlike SGK, SGKL is not induced by serum or glucocorticoids. It is induced in response to signals that activate phosphatidylinositol 3-kinase, which is also true for SGK. The protein phosphorylates several target proteins and has a role in neutral amino acid transport and activation of potassium and chloride channels. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            Bioinformatics

            Protein Aliases: Cytokine-independent survival kinase; DKFZp781N0293; Serine/threonine-protein kinase Sgk3; serum/glucocorticoid regulated kinase 3; Serum/glucocorticoid-regulated kinase 3; Serum/glucocorticoid-regulated kinase-like

            View more View less

            Gene Aliases: 2510015P22Rik; A330005P07Rik; CISK; fy; fz; SGK2; SGK3; SGKL

            View more View less

            UniProt ID: (Human) Q96BR1, (Mouse) Q9ERE3

            View more View less

            Entrez Gene ID: (Human) 23678, (Mouse) 170755, (Rat) 171498

            View more View less

            Function(s)

            protein kinase activity protein serine/threonine kinase activity calcium channel regulator activity protein binding ATP binding potassium channel regulator activity sodium channel regulator activity chloride channel regulator activity phosphatidylinositol binding nucleotide binding kinase activity transferase activity transferase activity, transferring phosphorus-containing groups non-receptor serine/threonine protein kinase

            Process(es)

            regulation of cell growth protein phosphorylation cellular sodium ion homeostasis response to stress positive regulation of sodium ion transport peptidyl-serine phosphorylation regulation of cell migration positive regulation of transporter activity ion transmembrane transport intracellular signal transduction regulation of cell proliferation regulation of apoptotic process regulation of sequence-specific DNA binding transcription factor activity phosphorylation negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            United States flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-584499df9d-6gk46:80/100.66.79.195:80.
            git-commit: 256cec658627a60c8e1196038bf5905a13cd45ba
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.33.0-2025.07.30-1.0