Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MAKERCLKKS FQDSLEDIKK RMKEKRNKNL AEIGKRRSFI AAPCQIITNT STLLKNYQDN NKMLVLALEN EKSKVKEAQD IILQLRKECY YLTCQLYALK GKLTSQQTVE PAQNQEICSS GMDPNSDDSS RNLFVKDLPQ IPLEETELPG QGESFQIEAT PPETQQSPHL SLKDITNVSL YPVVKIRRLS LSPKKNKASP AVALPKRRCT ASVNYKEPTL ASKLRRGDPF TDLCFLNSPI FKQKKDLRRS KKRALEVSPA KEAIFILYYV REFVSRFPDC RKCKLETHIC LR
The protein encoded by this gene is a member of the shugoshin family of proteins. This protein is thought to protect centromeric cohesin from cleavage during mitotic prophase by preventing phosphorylation of a cohesin subunit. Reduced expression of this gene leads to the premature loss of centromeric cohesion, mis-segregation of sister chromatids, and mitotic arrest. Evidence suggests that this protein also protects a small subset of cohesin found along the length of the chromosome arms during mitotic prophase. An isoform lacking exon 6 has been shown to play a role in the cohesion of centrioles. Mutations in this gene have been associated with Chronic Atrial and Intestinal Dysrhythmia (CAID) syndrome, characterized by the co-occurrence of Sick Sinus Syndrome (SSS) and Chronic Intestinal Pseudo-obstruction (CIPO) within the first four decades of life. Fibroblast cells from CAID patients exhibited both increased cell proliferation and higher rates of senescence. Pseudogenes of this gene have been found on chromosomes 1 and 7. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hSgo1; Serologically defined breast cancer antigen NY-BR-85; Shugoshin 1; shugoshin 1AB protein; shugoshin 1CD protein; shugoshin 1EF protein; shugoshin 1GH protein; shugoshin 1KL protein; Shugoshin-like 1; shugoshin-like 1 (S. pombe)
Gene Aliases: CAID; NY-BR-85; SGO; SGO1; SGOL1
UniProt ID: (Human) Q5FBB7
Entrez Gene ID: (Human) 151648, (Rat) 363174
Molecular Function:
centromere DNA-binding protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support