Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
The recommended shelf life for this product is 1 year from date of receipt.
May play a cooperative role in NF2-mediated growth suppression of cells. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cx43-interacting protein of 85-kDa; DKFZp761D051; merlin binding protein; Merlin-associated protein; Rab-GTPase-activating protein-like protein; RabGAPLP; RUN and SH3 containing 3; RUN and TBC1 domain containing 3; RUN and TBC1 domain-containing protein 3; SGSM3; Small G protein signaling modulator 3; small G protein signaling modulator 3 protein
Gene Aliases: 1810012I01Rik; AI428557; BB175482; Cip85; MAP; R75178; RabGAP-5; RABGAP5; RABGAPLP; RUSC3; RUTBC3; SGSM3
UniProt ID: (Human) Q96HU1, (Mouse) Q8VCZ6
Entrez Gene ID: (Human) 27352, (Mouse) 105835
Molecular Function:
scaffold/adaptor protein
GTPase-activating protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support