Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MQVPQDGED LAGQPWYHGL LSRQKAEALL QQNGDFLVRA SGSRGGNPVI SCRWRGSALH FEVFRVALRP RPGRPTALFQ LEDEQFPSIP ALVHSYMTGR RPLSQATGAV VSRPVTWQGP LRRSFSEDTL MDGPARIEPL RARKWSNSQP ADLAHMGRSR EDPAGMEAST MPISALPRTS SDPVLLKAPA PLGTVADSLR ASDGQLQAKA PTKPPRTPSF ELPDASERPP TYCELVPRVP SVQGTSPSQS CPEPEAPWWE AEEDEEEENR CFTRPQAEIS FCPHDAPSCL LGPQNRPLEP QVLHTLRGLF LEHHPGSTAL HLLLVDCQAT GLLGVTRDQR GNMGVSSGLE L (1-350 aa encoded by BC006281)
May play a role in JNK activation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Novel SH2-containing protein 1; SH2 domain-containing 3A; SH2 domain-containing protein 3A
Gene Aliases: NSP1; SH2D3A; UNQ175/PRO201
UniProt ID: (Human) Q9BRG2
Entrez Gene ID: (Human) 10045
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support